3-Hydroxybenzoic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
m-Hydroxybenzoic Acid
H0205-25G
H0205-500G
H0205-100G
TCH0205-025GEA
54.87
CAD
TCH0205-025G
TCH0205-500G
TCH0205-100G
3-Hydroxybenzoic acid ≥98.0%
3-Hydroxybenzoic acid
CAS Number: 99-06-9
MDL Number: MFCD00002506
Molecular Formula: C7H6O3
Molecular Weight: 138.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 203
Formula:
C₇H₆O₃ MW: 138.12 g/mol Boiling Pt: 346 Melting Pt: 203 °C Density: 1.473 g/cm³ (20 °C) Flash Pt: 220 Storage Temperature: Ambient |
MDL Number:
MFCD00002506 CAS Number: 99-06-9 EINECS: 202-726-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...