DL-Homocysteine 90,0

Supplier: TCI America

Synonyms: (±)-Homocysteine, 2-Amino-4-mercaptobutanoic acid, H-DL-HoCys-OH

H0159-5G H0159-1G
TCH0159-5GEA 364.79 CAD
TCH0159-5G TCH0159-001G
DL-Homocysteine 90,0
DL-Homocysteine

CAS Number: 454-29-5
MDL Number: MFCD00004898
Molecular Formula: C4H9NO2S
Molecular Weight: 135.18
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Melting point (°C): 233
Flash Point (°C): 110

Formula: C₄H₉NO₂S
MW: 135.19 g/mol
Melting Pt: 233 °C
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00004898
CAS Number: 454-29-5
EINECS: 207-222-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=135b293a1a464559cb498af13eb5844d452982a2&vid=135b293a1a464559cb498af13eb5844d452982a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR