Butyl hexanoate ≥98.0%

Supplier: TCI America

H0107-25ML H0107-500ML
TCH0107-025MLEA 49.86 CAD
TCH0107-025ML TCH0107-500ML
Butyl hexanoate ≥98.0%
Butyl hexanoate

CAS Number: 626-82-4
MDL Number: MFCD00053804
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 102
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Hexanoic Acid Butyl Ester

Formula: C₁₀H₂₀O₂
Boiling Pt: 208 °C
Density: 0.87 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00053804
CAS Number: 626-82-4
EINECS: 210-964-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=191debaa3449aabf11ef86f6e4489fd4aba1284d&vid=191debaa3449aabf11ef86f6e4489fd4aba1284d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR