Heptadecylbenzene ≥98.0%

Supplier: TCI America

Synonyms: 1-Phenylheptadecane

H0020-5G H0020-25G
TCH0020-005GEA 104.28 CAD
TCH0020-005G TCH0020-025G
Heptadecylbenzene ≥98.0%
Heptadecylbenzene

CAS Number: 14752-75-1
MDL Number: MFCD00039873
Molecular Formula: C23H40
Molecular Weight: 316.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Other Product Names / Synonyms: 1-Phenylheptadecane

Formula: C₂₃H₄₀
Boiling Pt: 397 °C (1013 hPa)
Melting Pt: 32 °C
Density: 0.8546 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039873
CAS Number: 14752-75-1
EINECS: 238-812-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5adf56d20ef08c55a9028d05b18b083cdcdbc1fd&vid=cfc5d5e425fd3099ff9d45347388cc0e1601f72f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR