Heneicosanoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

H0010-1G H0010-10G
TCH0010-1GEA 257.54 CAD
TCH0010-1G TCH0010-010G
Heneicosanoic acid ≥98.0% (by GC, titration analysis)
Heneicosanoic acid

CAS Number: 2363-71-5
MDL Number: MFCD00002805
Molecular Formula: C21H42O2
Molecular Weight: 326.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 171
Melting point (°C): 75
Flash Point (°C): 360

Formula: C₂₁H₄₂O₂
Boiling Pt: 171 °C
Melting Pt: 75 °C
Flash Pt: 360 °C
Storage Temperature: Ambient
MDL Number: MFCD00002805
CAS Number: 2363-71-5
EINECS: 219-113-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a7ee081f27ef92bd110623b69654717c8572c76&vid=adda898afa791c84b1ae6e8a1e4b6295b1ffc5f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR