4-Guanidinobenzoic acid hydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

G0238-25G G0238-5G
TCG0238-025GEA 83.74 CAD
TCG0238-025G TCG0238-005G
4-Guanidinobenzoic acid hydrochloride ≥97.0% (by titrimetric analysis)
4-Guanidinobenzoic acid hydrochloride

CAS Number: 42823-46-1
MDL Number: MFCD00040587
Molecular Formula: C8H9N3O2
Molecular Weight: 215.64
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 276

Formula: C₈H₁₀ClN₃O₂
Melting Pt: 276 °C
MDL Number: MFCD00040587
CAS Number: 42823-46-1
EINECS: 255-956-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab2c713c1b0d55a90feb2a01e8d65508266698db&vid=ab2c713c1b0d55a90feb2a01e8d65508266698db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR