Trilaurin (glycerol trilaurate)

Supplier: TCI America

Synonyms: Glyceryl trilaurate, Glycerol tridodecanoate, Glyceryl tridodecanoate, Glycerol trilaurate

G0087-25G G0087-500G G0087-100G
TCG0087-025GEA 91.64 CAD
TCG0087-025G TCG0087-500G TCG0087-100G
Trilaurin (glycerol trilaurate)
Trilaurin (Glycerol trilaurate)

CAS Number: 538-24-9
MDL Number: MFCD00026559
Molecular Formula: C39H74O6
Molecular Weight: 639.02
Form: Crystal
Color: White
Melting point (°C): 47


Other Product Names / Synonyms: Glycerol Trilaurate

Formula: C₃₉H₇₄O₆
MW: 639.01 g/mol
Boiling Pt: 643
Melting Pt: 49
Density: 0.9713 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00026559
CAS Number: 538-24-9
EINECS: 208-687-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=411710aa57dc9b6b66b597109d38695c10f5f700&vid=411710aa57dc9b6b66b597109d38695c10f5f700&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR