5-Fluorouridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

F0636-5G F0636-1G
TCF0636-5GEA 237 CAD
TCF0636-5G TCF0636-1G
5-Fluorouridine ≥98.0% (by titrimetric analysis)
5-Fluorouridine

CAS Number: 316-46-1
MDL Number: MFCD00036832
Molecular Formula: C9H11FN2O6
Molecular Weight: 262.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 185
Specific rotation [a]20/D: 18 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: FUrd, 1-(beta-D-Ribofuranosyl)-5-fluorouracil

Formula: C₉H₁₁FN₂O₆
Melting Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00036832
CAS Number: 316-46-1
EINECS: 206-260-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9dbd8e48e16e98fe68dfa9024c4bbb1d4ca8633&vid=e9dbd8e48e16e98fe68dfa9024c4bbb1d4ca8633&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR