4-Fluoro-2-nitroanisole ≥98.0%

Supplier: TCI America
Warning

F0615-5G F0615-25G
TCF0615-5GEA 128.93 CAD
TCF0615-5G TCF0615-25G
4-Fluoro-2-nitroanisole ≥98.0%
4-Fluoro-2-nitroanisole

CAS Number: 445-83-0
MDL Number: MFCD00013375
Molecular Formula: C7H6FNO3
Molecular Weight: 171.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-1-methoxy-2-nitrobenzene

Formula: C₇H₆FNO₃
Melting Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00013375
CAS Number: 445-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56b0c20649aa5b8dcda4162f8733cb2e2d557035&vid=56b0c20649aa5b8dcda4162f8733cb2e2d557035&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR