3-Fluoroanthranilic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Amino-3-fluorobenzoic acid

F0570-5G F0570-1G
TCF0570-5GEA 170.64 CAD
TCF0570-5G TCF0570-1G
3-Fluoroanthranilic acid ≥98.0% (by titrimetric analysis)
3-Fluoroanthranilic acid

CAS Number: 825-22-9
MDL Number: MFCD01569395
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 187


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-3-fluorobenzoic Acid

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 187 °C
MDL Number: MFCD01569395
CAS Number: 825-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=171d503dcbfb1f9f97b5dc6c8b292ba9d03c4171&vid=171d503dcbfb1f9f97b5dc6c8b292ba9d03c4171&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR