2-Fluorothioanisole ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1-Fluoro-2-(methylthio)benzene, 2-Fluorophenyl methyl sulfide, 2-Fluorophenyl methyl sulphide
CAS Number: 655-20-9
MDL Number: MFCD03093762
Molecular Formula: C7H7FS
Molecular Weight: 142.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 194
Flash Point (°C): 91
Specific Gravity (20/20): 1.17
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluorophenyl Methyl Sulfide, 1-Fluoro-2-(methylthio)benzene
Formula:
C₇H₇FS Boiling Pt: 194 °C Density: 1.17 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD03093762 CAS Number: 655-20-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...