4-Fluoro-3-nitrotoluene ≥98.0%
Supplier: TCI America
F0562-25G
F0562-5G
TCF0562-25GEA
205.4
CAD
TCF0562-25G
TCF0562-5G
4-Fluoro-3-nitrotoluene ≥98.0%
4-Fluoro-3-nitrotoluene
CAS Number: 446-11-7
MDL Number: MFCD00007060
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 241
Freezing point (°C): 22
Formula:
C₇H₆FNO₂ Boiling Pt: 241 °C |
MDL Number:
MFCD00007060 CAS Number: 446-11-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...