1-Fluoroadamantane ≥98.0%

Supplier: TCI America

Synonyms: 1-Fluorotricyclo[3.3.1.1(3,7)]decane

F0511-250MG F0511-1G
TCF0511-250MGEA 140.62 CAD
TCF0511-250MG TCF0511-1G
1-Fluoroadamantane ≥98.0%
1-Fluoroadamantane

CAS Number: 768-92-3
Molecular Formula: C10H15F
Molecular Weight: 154.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 257


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Adamantyl Fluoride

Formula: C₁₀H₁₅F
Melting Pt: 257 °C
CAS Number: 768-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eed2ebc2ac162e4b75ec0a2c53a6f58b6ff34cf2&vid=eed2ebc2ac162e4b75ec0a2c53a6f58b6ff34cf2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR