5-Fluoro-2-methoxybenzonitrile ≥98.0%

Supplier: TCI America

F0479-1G F0479-5G
TCF0479-1GEA 63.2 CAD
TCF0479-1G TCF0479-5G
5-Fluoro-2-methoxybenzonitrile ≥98.0%
5-Fluoro-2-methoxybenzonitrile

CAS Number: 189628-38-4
MDL Number: MFCD02683503
Molecular Formula: C8H6FNO
Molecular Weight: 151.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Cyano-4-fluoroanisole

Formula: C₈H₆FNO
MDL Number: MFCD02683503
CAS Number: 189628-38-4
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06b0c9842a950d510ff97c30c98138a7de122ca4&vid=06b0c9842a950d510ff97c30c98138a7de122ca4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR