4-Fluorochalcone ≥98.0%

Supplier: TCI America

Synonyms: 4-Fluorobenzylidenacetophenone, 3-(4-Fluorophenyl)-1-phenylprop-2-en-1-one

F0459-25G F0459-5G
TCF0459-25GEA 195.92 CAD
TCF0459-25G TCF0459-5G
4-Fluorochalcone ≥98.0%
4-Fluorochalcone

CAS Number: 1608-51-1
MDL Number: MFCD00443467
Molecular Formula: C15H11FO
Molecular Weight: 226.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 87


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(4-Fluorobenzal)acetophenone, 2-(4-Fluorobenzylidene)acetophenone

Formula: C₁₅H₁₁FO
MW: 226.25 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00443467
CAS Number: 1608-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df48743891ddd87e5b0f006af9e4debc83cf9df2&vid=df48743891ddd87e5b0f006af9e4debc83cf9df2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR