5-Fluoroorotic acid monohydrate (5-FOA) ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

F0382-100MG F0382-1G
TCF0382-100MGEA 26.86 CAD
TCF0382-100MG TCF0382-001G
5-Fluoroorotic acid monohydrate (5-FOA) ≥95.0% (by HPLC, titration analysis)
5-Fluoroorotic acid monohydrate

CAS Number: 703-95-7
MDL Number: MFCD00150658
Molecular Formula: C5H3FN2O4
Molecular Weight: 174.09
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 5-Fluorouracil-6-carboxylic Acid

Formula: C₅H₃FN₂O₄·1H₂O
MDL Number: MFCD00150658
CAS Number: 220141-70-8
EINECS: 211-876-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c769e34212d77320c9cafd1aecbd0a31225f7fe&vid=6c769e34212d77320c9cafd1aecbd0a31225f7fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR