1-Ethyl-3-methylimidazolium iodide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
E0556-25G
E0556-5G
TCE0556-25GEA
131.14
CAD
TCE0556-25G
TCE0556-5G
1-Ethyl-3-methylimidazolium iodide ≥98.0% (by titrimetric analysis)
1-Ethyl-3-methylimidazolium iodide
CAS Number: 35935-34-3
MDL Number: MFCD03701101
Molecular Formula: C6H11IN2
Molecular Weight: 238.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 79
Formula:
C₆H₁₁IN₂ Melting Pt: 79 °C |
MDL Number:
MFCD03701101 CAS Number: 35935-34-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...