1-Ethylpyridinium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

E0544-25G E0544-5G
TCE0544-25GEA 576.7 CAD
TCE0544-25G TCE0544-5G
1-Ethylpyridinium chloride ≥98.0% (by titrimetric analysis)
1-Ethylpyridinium chloride

CAS Number: 2294-38-4
MDL Number: MFCD03093293
Molecular Formula: C7H10ClN
Molecular Weight: 143.61
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₇H₁₀ClN
MDL Number: MFCD03093293
CAS Number: 2294-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d2c0560b41494c0d2126cba5cf55c0fdcb33676&vid=3d2c0560b41494c0d2126cba5cf55c0fdcb33676&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR