1-Ethoxynaphthalene ≥95.0%

Supplier: TCI America

E0532-25G E0532-5G
TCE0532-25GEA 327.06 CAD
TCE0532-25G TCE0532-5G
1-Ethoxynaphthalene ≥95.0%
1-Ethoxynaphthalene

CAS Number: 5328-01-8
MDL Number: MFCD00003928
Molecular Formula: C12H12O
Molecular Weight: 172.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 112
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Ethyl 1-Naphthyl Ether

Formula: C₁₂H₁₂O
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 5.5 °C
Density: 1.06 (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00003928
CAS Number: 5328-01-8
EINECS: 226-213-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dc160387f2ac729cde110bcfa193cf80aeccfec&vid=5dc160387f2ac729cde110bcfa193cf80aeccfec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR