4-Ethynylaniline ≥98.0%

Supplier: TCI America

Synonyms: (4-Aminophenyl)acetylene, 1-Ethynyl-4-aminobenzene, 1-Amino-4-ethynylbenzene, (4-Aminophenyl)ethyne

E0505-10G E0505-25G
TCE0505-010GEA 404.48 CAD
TCE0505-010G TCE0505-25G
4-Ethynylaniline ≥98.0%
4-Ethynylaniline

CAS Number: 14235-81-5
MDL Number: MFCD00168824
Molecular Formula: C8H7N
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 104


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Aminophenylacetylene

Formula: C₈H₇N
MW: 117.15 g/mol
Melting Pt: 104 °C
MDL Number: MFCD00168824
CAS Number: 14235-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fefed09a1d4acb2275b99710143c1941c5decca7&vid=fefed09a1d4acb2275b99710143c1941c5decca7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR