2-Hydroxyethyl salicylate ≥98.0%
Supplier: TCI America
E0113-500G
E0113-25G
TCE0113-500GEA
197.5
CAD
TCE0113-500G
TCE0113-025G
2-Hydroxyethyl salicylate ≥98.0%
2-Hydroxyethyl salicylate
CAS Number: 87-28-5
MDL Number: MFCD00002862
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 172
Flash Point (°C): 174
Freezing point (°C): 26
Other Product Names / Synonyms: Glycol Monosalicylate, Ethylene Glycol Monosalicylate, Salicylic Acid 2-Hydroxyethyl Ester
Formula:
C₉H₁₀O₄ MW: 182.18 g/mol Boiling Pt: 172 °C Flash Pt: 174 °C |
MDL Number:
MFCD00002862 CAS Number: 87-28-5 EINECS: 201-737-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...