2-(N-Ethylanilino)ethanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0062-25G E0062-500G
TCE0062-025GEA 66.68 CAD
TCE0062-025G TCE0062-500G
2-(N-Ethylanilino)ethanol ≥98.0% (by GC, titration analysis)
2-(N-Ethylanilino)ethanol

CAS Number: 92-50-2
MDL Number: MFCD00020575
Molecular Formula: C10H15NO
Molecular Weight: 165.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 268
Flash Point (°C): 132
Freezing point (°C): 34


Other Product Names / Synonyms: N-Ethyl-N-2-ethanolaniline, N-Ethyl-N-(2-hydroxyethyl)aniline, Ethylphenylethanolamine

Formula: C₁₀H₁₅NO
Boiling Pt: 268 °C
Flash Pt: 132 °C
MDL Number: MFCD00020575
CAS Number: 92-50-2
EINECS: 202-160-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcb5397e8cdcce19715c551459004399848562e1&vid=bcb5397e8cdcce19715c551459004399848562e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR