Arachidic acid ≥98.0%

Supplier: TCI America

Synonyms: Eicosanylic acid, Arachidinic acid, Icosanoic acid, Eicosanoic acid

E0006-25G E0006-1G
TCE0006-025GEA 211.72 CAD
TCE0006-025G TCE0006-001G
Arachidic acid ≥98.0%
Arachidic acid

CAS Number: 506-30-9
MDL Number: MFCD00002755
Molecular Formula: C20H40O2
Molecular Weight: 312.54
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 303
Melting point (°C): 76
Flash Point (°C): 110


Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Eicosanoic Acid, Icosanoic Acid

Formula: C₂₀H₄₀O₂
Boiling Pt: 303 °C
Melting Pt: 76 °C
Density: 0.824 g/cm³ (100 °C) (lit.)
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00002755
CAS Number: 506-30-9
EINECS: 208-031-3
Merck Index: 12,00802
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d33bd761fc1b0388163d10611a9598ce58d40dc&vid=9e6c92b69ad734eeae45abe77f1c749146f76d52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR