1,2-Dimethyl-3-propylimidazolium iodide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1,2-Dimethyl-3-propyl-1H-imidazol-3-ium iodide

D3903-25G D3903-5G
TCD3903-25GEA 500.86 CAD
TCD3903-25G TCD3903-5G
1,2-Dimethyl-3-propylimidazolium iodide ≥98.0% (by HPLC, titration analysis)
1,2-Dimethyl-3-propylimidazolium iodide

CAS Number: 218151-78-1
Molecular Formula: C8H15IN2
Molecular Weight: 266.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₈H₁₅IN₂
MW: 266.1226 g/mol
MDL Number: MFCD15072160
CAS Number: 218151-78-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ff1eb727c84ea8fcd9287e72a5f90ec00133ad5&vid=9ff1eb727c84ea8fcd9287e72a5f90ec00133ad5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR