6-(Dimethylamino)purine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3894-5G D3894-1G
TCD3894-5GEA 1112.32 CAD
TCD3894-5G TCD3894-1G
6-(Dimethylamino)purine ≥98.0% (by HPLC, titration analysis)
6-(Dimethylamino)purine

CAS Number: 938-55-6
MDL Number: MFCD00005573
Molecular Formula: C7H9N5
Molecular Weight: 163.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 260


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: N6,N6-Dimethyladenine

Formula: C₇H₉N₅
Melting Pt: 260 °C
MDL Number: MFCD00005573
CAS Number: 938-55-6
EINECS: 213-344-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bddc59b665f295ed0f5cf542dcb51d33c9d6c3d0&vid=bddc59b665f295ed0f5cf542dcb51d33c9d6c3d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR