2,3-Dimethylhydroquinone ≥98.0%
Supplier: TCI America
D3877-25G
D3877-5G
TCD3877-25GEA
521.4
CAD
TCD3877-25G
TCD3877-5G
2,3-Dimethylhydroquinone ≥98.0%
2,3-Dimethylhydroquinone
CAS Number: 608-43-5
MDL Number: MFCD00009997
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 224
Other Product Names / Synonyms: 3,6-Dihydroxy-o-xylene
Formula:
C₈H₁₀O₂ Melting Pt: 224 °C |
MDL Number:
MFCD00009997 CAS Number: 608-43-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...