1,4-Dichlorophthalazine ≥98.0%

Supplier: TCI America

D3749-1G D3749-5G
TCD3749-1GEA 61.62 CAD
TCD3749-1G TCD3749-5G
1,4-Dichlorophthalazine ≥98.0%
1,4-Dichlorophthalazine

CAS Number: 4752-10-7
MDL Number: MFCD00006909
Molecular Formula: C8H4Cl2N2
Molecular Weight: 199.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 166

Formula: C₈H₄Cl₂N₂
Melting Pt: 166 °C
MDL Number: MFCD00006909
CAS Number: 4752-10-7
EINECS: 225-275-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec37d4c18990d2d7494d9dbf277d8aaad787e993&vid=ec37d4c18990d2d7494d9dbf277d8aaad787e993&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR