2,5-Dimethylindole ≥97.0%

Supplier: TCI America

D3698-5G D3698-1G
TCD3698-5GEA 378.57 CAD
TCD3698-5G TCD3698-1G
2,5-Dimethylindole ≥97.0%
2,5-Dimethylindole

CAS Number: 1196-79-8
MDL Number: MFCD00005621
Molecular Formula: C10H11N
Molecular Weight: 145.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 115

Formula: C₁₀H₁₁N
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00005621
CAS Number: 1196-79-8
EINECS: 214-816-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30a65b06d9f7620a630c7ef450a7fe5944fc9016&vid=30a65b06d9f7620a630c7ef450a7fe5944fc9016&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR