2,4-Dibromothiazole ≥98.0%
Supplier: TCI America
D3667-1G
D3667-25G
D3667-5G
TCD3667-1GEA
33.18
CAD
TCD3667-1G
TCD3667-25G
TCD3667-5G
2,4-Dibromothiazole ≥98.0%
2,4-Dibromothiazole
CAS Number: 4175-77-3
MDL Number: MFCD01318994
Molecular Formula: C3HBr2NS
Molecular Weight: 242.92
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82
Formula:
C₃HBr₂NS Melting Pt: 82 °C |
MDL Number:
MFCD01318994 CAS Number: 4175-77-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...