2,7-Dibromonaphthalene ≥98.0%
Supplier: TCI America
D3624-25G
D3624-5G
TCD3624-25GEA
873.74
CAD
TCD3624-25G
TCD3624-5G
2,7-Dibromonaphthalene ≥98.0%
2,7-Dibromonaphthalene
CAS Number: 58556-75-5
MDL Number: MFCD00238580
Molecular Formula: C10H6Br2
Molecular Weight: 285.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 141
Formula:
C₁₀H₆Br₂ Melting Pt: 141 °C |
MDL Number:
MFCD00238580 CAS Number: 58556-75-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...