1,3-Dichloro-2-iodobenzene ≥98.0%
Supplier: TCI America
D3593-25G
D3593-5G
TCD3593-25GEA
167.48
CAD
TCD3593-25G
TCD3593-5G
1,3-Dichloro-2-iodobenzene ≥98.0%
1,3-Dichloro-2-iodobenzene
CAS Number: 19230-28-5
MDL Number: MFCD00060658
Molecular Formula: C6H3Cl2I
Molecular Weight: 272.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 64
Formula:
C₆H₃Cl₂I MW: 272.90 g/mol Melting Pt: 64 °C |
MDL Number:
MFCD00060658 CAS Number: 19230-28-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...