2,4-Dibromotoluene ≥98.0%

Supplier: TCI America

Synonyms: 2,4-Dibromo-1-methylbenzene

D3591-1G D3591-5G
TCD3591-1GEA 28.44 CAD
TCD3591-1G TCD3591-5G
2,4-Dibromotoluene ≥98.0%
2,4-Dibromotoluene

CAS Number: 31543-75-6
MDL Number: MFCD00052985
Molecular Formula: C7H6Br2
Molecular Weight: 249.93
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 243
Melting point (°C): -10
Specific Gravity (20/20): 1.85

Formula: C₇H₆Br₂
Boiling Pt: 243 °C
Melting Pt: –10 °C
Density: 1.85 (20 °C)
MDL Number: MFCD00052985
CAS Number: 31543-75-6
EINECS: 250-689-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR