2'-Deoxyinosine ≥98.0%

Supplier: TCI America

Synonyms: 9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-ol

D3584-1G D3584-5G
TCD3584-1GEA 127.35 CAD
TCD3584-1G TCD3584-5G
2'-Deoxyinosine ≥98.0%
2'-Deoxyinosine

CAS Number: 890-38-0
MDL Number: MFCD00005762
Molecular Formula: C10H12N4O4
Molecular Weight: 252.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 220
Specific rotation [a]20/D: -31 deg (C=1, 0.1mol/L NaOH)


Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: 9-(2-Deoxy-beta-D-ribofuranosyl)hypoxanthine

Formula: C₁₀H₁₂N₄O₄
Melting Pt: 220 °C
MDL Number: MFCD00005762
CAS Number: 890-38-0
EINECS: 212-964-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa8a87bb81b09ff31f7f511ab494c11f02a59ed7&vid=fa8a87bb81b09ff31f7f511ab494c11f02a59ed7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR