2'-Deoxycytidine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3583-5G D3583-1G
TCD3583-5GEA 158 CAD
TCD3583-5G TCD3583-1G
2'-Deoxycytidine ≥98.0% (by HPLC, titration analysis)
2'-Deoxycytidine

CAS Number: 951-77-9
MDL Number: MFCD00006547
Molecular Formula: C9H13N3O4
Molecular Weight: 227.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 204
Specific rotation [a]20/D: 59 deg (C=1, H2O)


Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: 1-(2-Deoxy-beta-D-ribofuranosyl)cytosine, dC

Formula: C₉H₁₃N₃O₄
Melting Pt: 204 °C
MDL Number: MFCD00006547
CAS Number: 951-77-9
EINECS: 213-454-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ee4cd46214109310caa1db6e79dfe60b5cd138b&vid=1ee4cd46214109310caa1db6e79dfe60b5cd138b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR