2,4-Dichloro-5-methoxypyrimidine ≥98.0%

Supplier: TCI America

D3508-25G D3508-5G
TCD3508-25GEA 559.32 CAD
TCD3508-25G TCD3508-5G
2,4-Dichloro-5-methoxypyrimidine ≥98.0%
2,4-Dichloro-5-methoxypyrimidine

CAS Number: 19646-07-2
MDL Number: MFCD02685670
Molecular Formula: C5H4Cl2N2O
Molecular Weight: 179.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68

Formula: C₅H₄Cl₂N₂O
Melting Pt: 68 °C
MDL Number: MFCD02685670
CAS Number: 19646-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abfbcc51c79ca597cfa93f31cba7e284783e72d2&vid=abfbcc51c79ca597cfa93f31cba7e284783e72d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR