2,4-Dihydroxybenzamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3460-25G D3460-5G
TCD3460-25GEA 120.08 CAD
TCD3460-25G TCD3460-5G
2,4-Dihydroxybenzamide ≥98.0% (by HPLC, titration analysis)
2,4-Dihydroxybenzamide

CAS Number: 3147-45-3
MDL Number: MFCD00064913
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 228

Formula: C₇H₇NO₃
Melting Pt: 228 °C
MDL Number: MFCD00064913
CAS Number: 3147-45-3
EINECS: 221-567-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2c1c803a98c5e89f02f32079c2f8f6c12ba08c3&vid=b2c1c803a98c5e89f02f32079c2f8f6c12ba08c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR