2-(2,4-Difluorophenyl)pyridine ≥98.0%

Supplier: TCI America

D3413-1G D3413-5G
TCD3413-1GEA 60.04 CAD
TCD3413-1G TCD3413-5G
2-(2,4-Difluorophenyl)pyridine ≥98.0%
2-(2,4-Difluorophenyl)pyridine

CAS Number: 391604-55-0
MDL Number: MFCD07787526
Molecular Formula: C11H7F2N
Molecular Weight: 191.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.26

Formula: C₁₁H₇F₂N
MW: 191.18 g/mol
Density: 1.26 (20 °C)
MDL Number: MFCD07787526
CAS Number: 391604-55-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0de09cf5f8be9532bf3a4ccd00529de4a280e936&vid=0de09cf5f8be9532bf3a4ccd00529de4a280e936&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR