2-(Dicyclohexylphosphino)biphenyl ≥98.0%

Supplier: TCI America

Synonyms: (2-Biphenyl)dicyclohexylphosphine

D3388-1G D3388-5G
TCD3388-1GEA 64.46 CAD
TCD3388-1G TCD3388-5G
2-(Dicyclohexylphosphino)biphenyl ≥98.0%
2-(Dicyclohexylphosphino)biphenyl

CAS Number: 247940-06-3
MDL Number: MFCD01862441
Molecular Formula: C24H31P
Molecular Weight: 350.49
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 104


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: (2-Biphenylyl)dicyclohexylphosphine, CyJohnPhos

Formula: C₂₄H₃₁P
MW: 350.48 g/mol
Melting Pt: 104 °C
MDL Number: MFCD01862441
CAS Number: 247940-06-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd98e34c39ae8f066ea1a257df81de6ff0061203&vid=fd98e34c39ae8f066ea1a257df81de6ff0061203&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR