2,4-Dichloro-5-fluoropyrimidine ≥98.0%
Supplier: TCI America
D3374-25G
D3374-5G
TCD3374-25GEA
304.91
CAD
TCD3374-25G
TCD3374-5G
2,4-Dichloro-5-fluoropyrimidine ≥98.0%
2,4-Dichloro-5-fluoropyrimidine
CAS Number: 2927-71-1
Molecular Formula: C4HCl2FN2
Molecular Weight: 166.96
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 80
Melting point (°C): 39
Flash Point (°C): 106
Storage Temperature: 0-10°C
Formula:
C₄HCl₂FN₂ Boiling Pt: 80 °C Melting Pt: 39 °C Flash Pt: 106 °C |
MDL Number:
MFCD00233551 CAS Number: 2927-71-1 UN: 3261 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...