3,5-Dibromoanisole ≥98.0%
Supplier: TCI America
Synonyms:
1,3-Dibromo-5-methoxybenzene
D3364-5G
D3364-25G
TCD3364-5GEA
71.1
CAD
TCD3364-5G
TCD3364-25G
3,5-Dibromoanisole ≥98.0%
3,5-Dibromoanisole
CAS Number: 74137-36-3
MDL Number: MFCD02258848
Molecular Formula: C7H6Br2O
Molecular Weight: 265.93
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 261
Melting point (°C): 39
Flash Point (°C): 110
Other Product Names / Synonyms: 1,3-Dibromo-5-methoxybenzene
Formula:
C₇H₆Br₂O MW: 265.93 g/mol Boiling Pt: 261 °C Melting Pt: 39 °C Flash Pt: 110 °C |
MDL Number:
MFCD02258848 CAS Number: 74137-36-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...