7-(Dimethylamino)-4-methylcoumarin ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D3355-5G D3355-25G
TCD3355-5GEA 198.29 CAD
TCD3355-5G TCD3355-25G
7-(Dimethylamino)-4-methylcoumarin ≥98.0% (by titrimetric analysis)
7-(Dimethylamino)-4-methylcoumarin

CAS Number: 87-01-4
Molecular Formula: C12H13NO2
Molecular Weight: 203.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 142

Formula: C₁₂H₁₃NO₂
Melting Pt: 142 °C
MDL Number: MFCD00006862
CAS Number: 87-01-4
EINECS: 201-717-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=882e2cc9afcbff3e12eb914f3da3828a7a957258&vid=882e2cc9afcbff3e12eb914f3da3828a7a957258&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR