1,3-Dimethylimidazolium chloride ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Dimethyl-1H-imidazol-3-ium chloride

D3341-5G D3341-25G
TCD3341-5GEA 330.22 CAD
TCD3341-5G TCD3341-25G
1,3-Dimethylimidazolium chloride ≥98.0%
1,3-Dimethylimidazolium chloride

CAS Number: 79917-88-7
Molecular Formula: C5H9ClN2
Molecular Weight: 132.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 125

Formula: C₅H₉ClN₂
Melting Pt: 125 °C
MDL Number: MFCD08276366
CAS Number: 79917-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=364c5b53227b588da767aa1bde8ec229dd9a94f6&vid=364c5b53227b588da767aa1bde8ec229dd9a94f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR