4-(Diphenylphosphino)benzoic acid 97,0

Supplier: TCI America

D3243-1G D3243-5G
TCD3243-1GEA 225.94 CAD
TCD3243-1G TCD3243-5G
4-(Diphenylphosphino)benzoic acid 97,0
4-(Diphenylphosphino)benzoic acid

CAS Number: 2129-31-9
Molecular Formula: C19H15O2P
Molecular Weight: 306.30
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 156

Formula: C₁₉H₁₅O₂P
MW: 306.3 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00407264
CAS Number: 2129-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79cd39d294c958b8f535f7371a5d5b9c71049aab&vid=79cd39d294c958b8f535f7371a5d5b9c71049aab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR