2,6-Dibromoanthracene ≥98.0% (by HPLC)
Supplier: TCI America
D3171-1G
D3171-5G
TCD3171-1GEA
556.16
CAD
TCD3171-1G
TCD3171-5G
2,6-Dibromoanthracene ≥98.0% (by HPLC)
2,6-Dibromoanthracene
CAS Number: 186517-01-1
Molecular Formula: C14H8Br2
Molecular Weight: 336.03
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Formula:
C₁₄H₈Br₂ MW: 336.0213 g/mol |
MDL Number:
MFCD08276341 CAS Number: 186517-01-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...