2',3'-Dideoxyinosine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3066-500MG D3066-100MG
TCD3066-500MGEA 210.14 CAD
TCD3066-500MG TCD3066-100MG
2',3'-Dideoxyinosine ≥98.0% (by HPLC, titration analysis)
2',3'-Dideoxyinosine

CAS Number: 69655-05-6
MDL Number: MFCD00077728
Molecular Formula: C10H12N4O3
Molecular Weight: 236.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 182
Specific rotation [a]20/D: -28 deg (C=0.34, H2O)


Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: ddI, 9-(2,3-Dideoxy-beta-D-ribofuranosyl)-6-oxopurine, Didanosine, 9-(2,3-Dideoxy-beta-D-ribofuranosyl)hypoxanthine

Formula: C₁₀H₁₂N₄O₃
Melting Pt: 182 °C
MDL Number: MFCD00077728
CAS Number: 69655-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3f0be1d32af53f8d1c64629ee00350fc8750f51&vid=c3f0be1d32af53f8d1c64629ee00350fc8750f51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR