2,6-Dibromo-4-(trifluoromethoxy)aniline ≥98.0%

Supplier: TCI America

D3038-25G D3038-5G
TCD3038-25GEA 222.78 CAD
TCD3038-25G TCD3038-5G
2,6-Dibromo-4-(trifluoromethoxy)aniline ≥98.0%
2,6-Dibromo-4-(trifluoromethoxy)aniline

CAS Number: 88149-49-9
MDL Number: MFCD00153113
Molecular Formula: C7H4Br2F3NO
Molecular Weight: 334.92
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-1,3-dibromo-5-(trifluoromethoxy)benzene

Formula: C₇H₄Br₂F₃NO
MW: 334.92 g/mol
MDL Number: MFCD00153113
CAS Number: 88149-49-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4249473c3cd41c2c3c12b9d7f5aa240a1daab229&vid=4249473c3cd41c2c3c12b9d7f5aa240a1daab229&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR