3,6-Dibromocarbazole ≥98.0%

Supplier: TCI America

D2983-1G D2983-25G D2983-5G
TCD2983-1GEA 33.18 CAD
TCD2983-1G TCD2983-25G TCD2983-5G
3,6-Dibromocarbazole ≥98.0%
3,6-Dibromocarbazole

CAS Number: 6825-20-3
Molecular Formula: C12H7Br2N
Molecular Weight: 325.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 212

Formula: C₁₂H₇Br₂N
Melting Pt: 212 °C
MDL Number: MFCD00004961
CAS Number: 6825-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68a9debbe011df9a689688285b713657af7f5678&vid=68a9debbe011df9a689688285b713657af7f5678&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR