3,7-Dithia-1,9-nonanediol ≥92.0%

Supplier: TCI America

Synonyms: 1,3-Bis(2-hydroxyethylthio)propane, 2,2'-(Trimethylenedithio)diethanol

D2845-25G D2845-5G
TCD2845-25GEA 641.32 CAD
TCD2845-25G TCD2845-5G
3,7-Dithia-1,9-nonanediol ≥92.0%
3,7-Dithia-1,9-nonanediol

CAS Number: 16260-48-3
MDL Number: MFCD01098524
Molecular Formula: C7H16O2S2
Molecular Weight: 196.32
Purity/Analysis Method: >92.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.18


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Bis(2-hydroxyethylthio)propane, 2,2'-(Trimethylenedithio)diethanol

Formula: C₇H₁₆O₂S₂
Density: 1.18 (20 °C)
MDL Number: MFCD01098524
CAS Number: 16260-48-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38a2671e8f47f0e56dd2dcbed9dc88da583191f4&vid=38a2671e8f47f0e56dd2dcbed9dc88da583191f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR