1,3-Diacetoxyacetone ≥98.0%

Supplier: TCI America

D2843-25G D2843-5G
TCD2843-25GEA 395 CAD
TCD2843-25G TCD2843-5G
1,3-Diacetoxyacetone ≥98.0%
1,3-Diacetoxyacetone

CAS Number: 6946-10-7
MDL Number: MFCD01310655
Molecular Formula: C7H10O5
Molecular Weight: 174.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 50


Other Product Names / Synonyms: 1,3-Diacetoxy-2-propanone

Formula: C₇H₁₀O₅
Melting Pt: 50 °C
MDL Number: MFCD01310655
CAS Number: 6946-10-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR