1,8-Dichlorohexadecafluorooctane ≥98.0%

Supplier: TCI America

Synonyms: 1,8-Dichloroperfluorooctane

D2804-5G D2804-25G
TCD2804-5GEA 71.1 CAD
TCD2804-5G TCD2804-25G
1,8-Dichlorohexadecafluorooctane ≥98.0%
1,8-Dichlorohexadecafluorooctane

CAS Number: 647-25-6
MDL Number: MFCD00155747
Molecular Formula: C8Cl2F16
Molecular Weight: 470.96
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,8-Dichloroperfluorooctane

Formula: C₈Cl₂F₁₆
MDL Number: MFCD00155747
CAS Number: 647-25-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8965e0aa78bd687eab2c14f177057a66e9837da8&vid=8965e0aa78bd687eab2c14f177057a66e9837da8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR